Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STX12
Entrez Gene ID
23673
Gene Name
syntaxin 12
Gene Aliases
STX13, STX14
Chromosome Location
Chr.1: 27773183 - 27824452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177424.2 NP_803173.1 4 - 5 548 104
GenBank mRNA AF123769.1 - 4 - 5 495 104
AK297585.1 - 4 - 5 476 104
AK299982.1 - 4 - 5 380 104
AK309990.1 - 3 - 4 251 104
AK315880.1 - 2 - 3 179 104
AL035306.1 - 4 - 5 491 104
BC046999.1 - 4 - 5 476 104
CR407620.1 - 4 - 5 402 104

View Full Product Details