The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190964.2 | NP_001177893.1 | 13 - 14 | 2485 | 113 |
NM_001190965.2 | NP_001177894.1 | 12 - 13 | 2500 | 113 | |
NM_003453.4 | NP_003444.1 | 13 - 14 | 2593 | 113 | |
NM_197968.3 | NP_932072.1 | 12 - 13 | 2546 | 113 | |
XM_005266520.3 | XP_005266577.1 | 12 - 13 | 2505 | 113 | |
XM_011535216.2 | XP_011533518.1 | 13 - 14 | 2175 | 113 | |
XM_011535222.1 | XP_011533524.1 | 12 - 13 | 2436 | 113 | |
XM_011535223.2 | XP_011533525.1 | 6 - 7 | 4890 | 113 | |
XM_011535224.2 | XP_011533526.1 | 6 - 7 | 791 | 113 | |
XM_017020728.1 | XP_016876217.1 | 12 - 13 | 2436 | 113 | |
XM_017020729.1 | XP_016876218.1 | 12 - 13 | 2411 | 113 | |
XM_017020731.1 | XP_016876220.1 | 13 - 14 | 2228 | 113 | |
XM_017020732.1 | XP_016876221.1 | 13 - 14 | 2250 | 113 | |
XM_017020733.1 | XP_016876222.1 | 14 - 15 | 2277 | 113 | |
XM_017020734.1 | XP_016876223.1 | 14 - 15 | 2263 | 113 | |
XM_017020735.1 | XP_016876224.1 | 13 - 14 | 2227 | 113 | |
XM_017020736.1 | XP_016876225.1 | 13 - 14 | 2399 | 113 | |
XM_017020737.1 | XP_016876226.1 | 13 - 14 | 2174 | 113 | |
XM_017020738.1 | XP_016876227.1 | 12 - 13 | 2489 | 113 | |
GenBank mRNA | AF012126.1 | - | 6 - 7 | 888 | 113 |
AF035374.1 | - | 10 - 11 | 1791 | 113 | |
AF060181.1 | - | 10 - 11 | 1843 | 113 | |
AJ224901.1 | - | 13 - 14 | 2479 | 113 | |
AK226118.1 | - | 12 - 13 | 2480 | 113 | |
AK289741.1 | - | 12 - 13 | 2471 | 113 | |
AK302917.1 | - | 7 - 8 | 860 | 113 | |
BC036372.1 | - | 12 - 13 | 2546 | 113 | |
BX647944.1 | - | 12 - 13 | 2500 | 113 | |
BX648905.1 | - | 5 - 6 | 2930 | 113 | |
Y13472.1 | - | 13 - 14 | 2491 | 113 |