Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM2
Entrez Gene ID
7750
Gene Name
zinc finger MYM-type containing 2
Gene Aliases
FIM, MYM, RAMP, SCLL, ZNF198
Chromosome Location
Chr.13: 19957412 - 20091845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190964.2 NP_001177893.1 24 - 25 4004 125
NM_001190965.2 NP_001177894.1 23 - 24 4019 125
NM_003453.4 NP_003444.1 24 - 25 4112 125
NM_197968.3 NP_932072.1 23 - 24 4065 125
XM_005266520.3 XP_005266577.1 23 - 24 4024 125
XM_011535216.2 XP_011533518.1 24 - 25 3694 125
XM_011535223.2 XP_011533525.1 17 - 18 6409 125
XM_011535224.2 XP_011533526.1 17 - 18 2310 125
XM_017020728.1 XP_016876217.1 23 - 24 3955 125
XM_017020729.1 XP_016876218.1 23 - 24 3930 125
XM_017020731.1 XP_016876220.1 24 - 25 3747 125
XM_017020732.1 XP_016876221.1 24 - 25 3769 125
XM_017020733.1 XP_016876222.1 25 - 26 3796 125
XM_017020734.1 XP_016876223.1 25 - 26 3782 125
XM_017020735.1 XP_016876224.1 24 - 25 3746 125
XM_017020736.1 XP_016876225.1 24 - 25 3918 125
XM_017020737.1 XP_016876226.1 24 - 25 3693 125
GenBank mRNA AF012126.1 - 17 - 18 2407 125
AF060181.1 - 21 - 22 3362 125
AJ224901.1 - 24 - 25 3998 125
AK226118.1 - 23 - 24 3999 125
BC036372.1 - 23 - 24 4065 125
BX647944.1 - 23 - 24 4019 125
BX648905.1 - 16 - 17 4449 125
Y13472.1 - 24 - 25 4010 125

View Full Product Details