Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZMYM2
- Entrez Gene ID
-
7750
- Gene Name
- zinc finger MYM-type containing 2
- Gene Aliases
-
FIM, MYM, RAMP, SCLL, ZNF198
- Chromosome Location
-
Chr.13: 19957412 - 20091845 on Build GRCh38
View Full Product Details