The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145031.2 | NP_001138503.1 | 8 - 9 | 1400 | 65 |
NM_001319191.1 | NP_001306120.1 | 8 - 9 | 1087 | 65 | |
NM_002658.4 | NP_002649.1 | 9 - 10 | 1115 | 65 | |
XM_011539866.2 | XP_011538168.1 | 9 - 10 | 1110 | 65 | |
GenBank mRNA | A18397.1 | - | 9 - 10 | 1065 | 65 |
A35395.1 | - | 9 - 10 | 1035 | 65 | |
AB208961.1 | - | 3 - 4 | 464 | 65 | |
AK297940.1 | - | 8 - 9 | 1056 | 65 | |
AK298560.1 | - | 8 - 9 | 1369 | 65 | |
AK312960.1 | - | 9 - 10 | 1084 | 65 | |
AY820134.1 | - | 7 - 8 | 906 | 65 | |
BC013575.2 | - | 9 - 10 | 1058 | 65 | |
BT007391.1 | - | 8 - 9 | 966 | 65 | |
D00244.1 | - | 9 - 10 | 1032 | 65 | |
D11143.1 | - | 7 - 8 | 906 | 65 | |
M15476.1 | - | 9 - 10 | 1042 | 65 | |
X02760.1 | - | 8 - 9 | 1006 | 65 | |
Z36790.1 | - | 1 - 2 | 54 | 65 |