The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291296.1 | NP_001278225.1 | 8 - 9 | 937 | 65 |
NM_007059.3 | NP_008990.2 | 10 - 11 | 1105 | 65 | |
NR_111923.1 | 11 - 12 | 1192 | 65 | ||
XM_011526399.2 | XP_011524701.1 | 13 - 14 | 2610 | 65 | |
XM_017026226.1 | XP_016881715.1 | 13 - 14 | 1455 | 65 | |
XM_017026227.1 | XP_016881716.1 | 12 - 13 | 1531 | 65 | |
XM_017026228.1 | XP_016881717.1 | 13 - 14 | 1455 | 65 | |
GenBank mRNA | AF105369.1 | - | 10 - 11 | 1037 | 65 |
AK023996.1 | - | 10 - 11 | 1060 | 65 | |
AK298672.1 | - | 11 - 12 | 1090 | 65 | |
BC009249.2 | - | 10 - 11 | 1020 | 65 |