The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291296.1 | NP_001278225.1 | 7 - 8 | 803 | 80 |
NM_007059.3 | NP_008990.2 | 9 - 10 | 971 | 80 | |
NR_111923.1 | 10 - 11 | 1058 | 80 | ||
XM_011526399.2 | XP_011524701.1 | 12 - 13 | 2476 | 80 | |
XM_017026226.1 | XP_016881715.1 | 12 - 13 | 1321 | 80 | |
XM_017026227.1 | XP_016881716.1 | 11 - 12 | 1397 | 80 | |
XM_017026228.1 | XP_016881717.1 | 12 - 13 | 1321 | 80 | |
GenBank mRNA | AF105369.1 | - | 9 - 10 | 903 | 80 |
AK023996.1 | - | 9 - 10 | 926 | 80 | |
AK225419.1 | - | 9 - 10 | 915 | 80 | |
AK298672.1 | - | 10 - 11 | 956 | 80 | |
BC009249.2 | - | 9 - 10 | 886 | 80 | |
DR759729.1 | - | 7 - 8 | 715 | 80 |