The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002516.3 | NP_002507.1 | 2 - 3 | 275 | 72 |
XM_006723230.3 | XP_006723293.1 | 3 - 4 | 327 | 72 | |
XM_017026838.1 | XP_016882327.1 | 2 - 3 | 551 | 72 | |
XM_017026839.1 | XP_016882328.1 | 2 - 3 | 377 | 72 | |
GenBank mRNA | AF083898.1 | - | 2 - 3 | 275 | 72 |
U70477.1 | - | 2 - 3 | 246 | 72 |