The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145303.2 | NP_001138775.2 | 14 - 15 | 2205 | 74 |
NM_144686.3 | NP_653287.2 | 14 - 15 | 2187 | 74 | |
XM_011526486.2 | XP_011524788.1 | 11 - 12 | 1690 | 74 | |
GenBank mRNA | AK314973.1 | - | 14 - 15 | 74 | |
AY236492.1 | - | 14 - 15 | 74 | ||
AY263166.1 | - | 14 - 15 | 74 | ||
BC025323.1 | - | 14 - 15 | 74 | ||
BC032477.1 | - | 8 - 9 | 74 | ||
BU680152.1 | - | 3 - 4 | 316 | 74 |