The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017666.4 | NP_060136.1 | 15 - 16 | 2186 | 151 |
XM_006724765.3 | XP_006724828.1 | 14 - 15 | 1990 | 151 | |
GenBank mRNA | AK290373.1 | - | 15 - 16 | 2137 | 151 |
AL834333.1 | - | 15 - 16 | 2117 | 151 | |
BC051728.1 | - | 15 - 16 | 2137 | 151 | |
DA441065.1 | - | 2 - 3 | 210 | 151 |