The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017666.4 | NP_060136.1 | 2 - 3 | 239 | 86 |
XM_006724765.3 | XP_006724828.1 | 2 - 3 | 190 | 86 | |
GenBank mRNA | AK000102.1 | - | 2 - 3 | 162 | 86 |
AK290373.1 | - | 2 - 3 | 190 | 86 | |
AL133204.1 | - | 2 - 3 | 174 | 86 | |
AL834333.1 | - | 2 - 3 | 170 | 86 | |
BC051728.1 | - | 2 - 3 | 190 | 86 | |
DR003059.1 | - | 2 - 3 | 238 | 86 |