The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006583.2 | NP_006574.1 | 4 - 5 | 580 | 125 |
GenBank mRNA | AF012270.1 | - | 4 - 5 | 596 | 125 |
BC128120.1 | - | 4 - 5 | 571 | 125 | |
BC128121.1 | - | 4 - 5 | 571 | 125 | |
BC128401.1 | - | 4 - 5 | 571 | 125 |