Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCKDK
Entrez Gene ID
10295
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
BCKDKD, BDK
Chromosome Location
Chr.16: 31108294 - 31117651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122957.2 NP_001116429.1 6 - 7 905 69
NM_001271926.1 NP_001258855.1 6 - 7 905 69
NM_005881.3 NP_005872.2 6 - 7 905 69
XM_017022859.1 XP_016878348.1 6 - 7 818 69
GenBank mRNA AF026548.1 - 6 - 7 815 69
AK130145.1 - 6 - 7 785 69
AK294691.1 - 5 - 6 614 69
BC007363.2 - 6 - 7 788 69
BC009872.2 - 6 - 7 768 69
CR542093.1 - 5 - 6 542 69

View Full Product Details