Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STIP1
Entrez Gene ID
10963
Gene Name
stress induced phosphoprotein 1
Gene Aliases
HEL-S-94n, HOP, IEF-SSP-3521, P60, STI1, STI1L
Chromosome Location
Chr.11: 64185272 - 64204548 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282652.1 NP_001269581.1 13 - 14 2251 82
NM_001282653.1 NP_001269582.1 13 - 14 1632 82
NM_006819.2 NP_006810.1 13 - 14 1704 82
GenBank mRNA AK291555.1 - 13 - 14 1631 82
BC002987.1 - 13 - 14 1619 82
BC039299.1 - 13 - 14 2251 82
BM679538.1 - 1 - 2 82
BT020010.1 - 12 - 13 1557 82
BT020011.1 - 12 - 13 1557 82
CR536512.1 - 12 - 13 1557 82
FJ224350.1 - 13 - 14 1619 82
M86752.1 - 13 - 14 1619 82

View Full Product Details