Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM25
Entrez Gene ID
58517
Gene Name
RNA binding motif protein 25
Gene Aliases
NET52, RED120, RNPC7, S164, Snu71, fSAP94
Chromosome Location
Chr.14: 73058436 - 73121675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021239.2 NP_067062.1 12 - 13 1676 152
XM_011537044.2 XP_011535346.1 13 - 14 1778 152
XM_011537045.2 XP_011535347.1 12 - 13 1588 152
GenBank mRNA AK094697.1 - 11 - 12 1520 152
AK125513.1 - 3 - 4 3141 152
AL832314.1 - 11 - 12 5092 152
BC113389.1 - 11 - 12 1456 152
BC113391.1 - 11 - 12 1456 152
BC136775.1 - 12 - 13 1513 152
BC136776.1 - 12 - 13 1513 152
BC144407.1 - 13 - 14 1581 152
BX647116.1 - 12 - 13 1629 152

View Full Product Details