Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V1C2
Entrez Gene ID
245973
Gene Name
ATPase H+ transporting V1 subunit C2
Gene Aliases
ATP6C2, VMA5
Chromosome Location
Chr.2: 10720974 - 10785110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039362.1 NP_001034451.1 4 - 5 396 107
NM_144583.3 NP_653184.2 4 - 5 396 107
XM_017003745.1 XP_016859234.1 4 - 5 385 107
GenBank mRNA AK292952.1 - 4 - 5 389 107
AY039759.1 - 3 - 4 308 107
BC012142.1 - 3 - 4 307 107
BX358022.2 - 4 - 5 396 107
CD636943.1 - 4 - 5 350 107
DB230703.1 - 4 - 5 389 107

View Full Product Details