The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039362.1 | NP_001034451.1 | 5 - 6 | 483 | 68 |
NM_144583.3 | NP_653184.2 | 5 - 6 | 483 | 68 | |
XM_011510339.2 | XP_011508641.1 | 6 - 7 | 480 | 68 | |
XM_011510340.2 | XP_011508642.1 | 6 - 7 | 480 | 68 | |
XM_011510341.2 | XP_011508643.1 | 6 - 7 | 480 | 68 | |
XM_017003745.1 | XP_016859234.1 | 5 - 6 | 472 | 68 | |
GenBank mRNA | AK292952.1 | - | 5 - 6 | 476 | 68 |
AY039759.1 | - | 4 - 5 | 395 | 68 | |
BC012142.1 | - | 4 - 5 | 394 | 68 | |
BX358022.2 | - | 5 - 6 | 483 | 68 | |
CD636943.1 | - | 5 - 6 | 437 | 68 | |
DB230703.1 | - | 5 - 6 | 476 | 68 |