Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V1C2
Entrez Gene ID
245973
Gene Name
ATPase H+ transporting V1 subunit C2
Gene Aliases
ATP6C2, VMA5
Chromosome Location
Chr.2: 10720974 - 10785110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144583.3 NP_653184.2 10 - 11 930 103
XM_011510340.2 XP_011508642.1 11 - 12 927 103
GenBank mRNA AK292952.1 - 10 - 11 923 103
AY039759.1 - 9 - 10 842 103
BC012142.1 - 9 - 10 841 103

View Full Product Details