The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171080.1 | NP_001164551.1 | 6 - 7 | 1125 | 80 |
NM_001324242.1 | NP_001311171.1 | 7 - 8 | 1197 | 80 | |
NM_001324243.1 | NP_001311172.1 | 7 - 8 | 1290 | 80 | |
NM_001324244.1 | NP_001311173.1 | 7 - 8 | 1197 | 80 | |
NM_018301.4 | NP_060771.3 | 6 - 7 | 1125 | 80 | |
NR_136728.1 | 6 - 7 | 793 | 80 | ||
NR_136729.1 | 7 - 8 | 887 | 80 | ||
NR_136730.1 | 7 - 8 | 1921 | 80 | ||
NR_136731.1 | 7 - 8 | 991 | 80 | ||
NR_136732.1 | 6 - 7 | 886 | 80 | ||
XM_011530980.2 | XP_011529282.1 | 7 - 8 | 1289 | 80 | |
XM_011530981.2 | XP_011529283.1 | 6 - 7 | 1217 | 80 | |
GenBank mRNA | AK001878.1 | - | 6 - 7 | 1098 | 80 |
AK023732.1 | - | 7 - 8 | 1916 | 80 | |
BC006986.1 | - | 6 - 7 | 80 |