The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002269.2 | NP_002260.2 | 8 - 9 | 894 | 93 |
XM_011535805.1 | XP_011534107.1 | 9 - 10 | 1072 | 93 | |
XM_017010838.1 | XP_016866327.1 | 9 - 10 | 1072 | 93 | |
XM_017010839.1 | XP_016866328.1 | 8 - 9 | 875 | 93 | |
XM_017010840.1 | XP_016866329.1 | 9 - 10 | 1032 | 93 | |
XM_017010841.1 | XP_016866330.1 | 5 - 6 | 455 | 93 | |
XM_017010842.1 | XP_016866331.1 | 7 - 8 | 786 | 93 | |
XM_017010843.1 | XP_016866332.1 | 8 - 9 | 910 | 93 | |
GenBank mRNA | AF005361.1 | - | 7 - 8 | 759 | 93 |
AK314206.1 | - | 8 - 9 | 887 | 93 | |
BC047409.1 | - | 8 - 9 | 886 | 93 |