The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278439.1 | NP_001265368.1 | 17 - 18 | 1789 | 85 |
NM_007056.2 | NP_008987.2 | 18 - 19 | 1975 | 85 | |
NR_103529.1 | 18 - 19 | 1979 | 85 | ||
GenBank mRNA | AF042800.1 | - | 18 - 19 | 1975 | 85 |
AK094681.1 | - | 16 - 17 | 2512 | 85 | |
AK293333.1 | - | 17 - 18 | 1785 | 85 | |
AY358944.1 | - | 18 - 19 | 1960 | 85 | |
BC013178.1 | - | 13 - 14 | 1261 | 85 | |
BC080554.1 | - | 17 - 18 | 1915 | 85 |