The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007056.2 | NP_008987.2 | 4 - 5 | 399 | 67 |
NR_103529.1 | 4 - 5 | 399 | 67 | ||
XM_011526396.2 | XP_011524698.1 | 4 - 5 | 394 | 67 | |
XM_011526397.2 | XP_011524699.1 | 3 - 4 | 244 | 67 | |
GenBank mRNA | AF042800.1 | - | 4 - 5 | 399 | 67 |
AK094681.1 | - | 3 - 4 | 993 | 67 | |
AY358944.1 | - | 4 - 5 | 380 | 67 | |
BC013178.1 | - | 4 - 5 | 344 | 67 | |
BC080554.1 | - | 3 - 4 | 339 | 67 | |
BC131496.1 | - | 3 - 4 | 257 | 67 |