The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001756.3 | NP_001747.2 | 3 - 4 | 983 | 79 |
GenBank mRNA | AK225591.1 | - | 4 - 5 | 893 | 79 |
AK290821.1 | - | 3 - 4 | 968 | 79 | |
AK313760.1 | - | 3 - 4 | 968 | 79 | |
BC017462.1 | - | 3 - 4 | 937 | 79 | |
BC056259.1 | - | 3 - 4 | 958 | 79 | |
BC058021.1 | - | 3 - 4 | 967 | 79 | |
J02943.1 | - | 2 - 3 | 914 | 79 |