Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD11B1
Entrez Gene ID
3290
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases
11-DH, 11-beta-HSD1, CORTRD2, HDL, HSD11, HSD11B, HSD11L, SDR26C1
Chromosome Location
Chr.1: 209686180 - 209734950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206741.1 NP_001193670.1 4 - 5 479 73
NM_005525.3 NP_005516.1 3 - 4 477 73
NM_181755.2 NP_861420.1 4 - 5 457 73
GenBank mRNA AK313973.1 - 3 - 4 419 73
BC012593.1 - 3 - 4 426 73
BG619051.1 - 4 - 5 451 73
BX345837.2 - 3 - 4 390 73
CF596989.1 - 3 - 4 479 73
CF597174.1 - 4 - 5 460 73
KJ534869.1 - 3 - 4 325 73

View Full Product Details