The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206741.1 | NP_001193670.1 | 6 - 7 | 814 | 132 |
NM_005525.3 | NP_005516.1 | 5 - 6 | 812 | 132 | |
NM_181755.2 | NP_861420.1 | 6 - 7 | 792 | 132 | |
GenBank mRNA | AK313973.1 | - | 5 - 6 | 754 | 132 |
BC012593.1 | - | 5 - 6 | 761 | 132 | |
KJ534869.1 | - | 5 - 6 | 660 | 132 |