Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
Chromosome Location
Chr.2: 64524305 - 64593005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002243.2 NP_001002243.1 3 - 4 2424 84
NM_017657.4 NP_060127.3 3 - 4 2424 84
NM_203437.3 NP_982261.2 3 - 4 2424 84
XM_005264380.1 XP_005264437.1 3 - 4 2424 84
GenBank mRNA AK000087.1 - 3 - 4 2399 84
AK021899.1 - 2 - 3 1047 84
AK222821.1 - 3 - 4 2347 84
AL833962.2 - 3 - 4 2202 84
AY367088.1 - 2 - 3 2084 84
BC047529.1 - 3 - 4 2398 84

View Full Product Details