Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
Chromosome Location
Chr.2: 64524305 - 64593005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017657.4 NP_060127.3 8 - 9 2917 93
XM_005264380.1 XP_005264437.1 9 - 10 3001 93
GenBank mRNA AK000087.1 - 8 - 9 2892 93
AK021899.1 - 8 - 9 1624 93
AK024658.1 - 6 - 7 553 93
AL833962.2 - 8 - 9 4422 93
BC047529.1 - 8 - 9 2891 93

View Full Product Details