Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIF1
Entrez Gene ID
11197
Gene Name
WNT inhibitory factor 1
Gene Aliases
WIF-1
Chromosome Location
Chr.12: 65050624 - 65121566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007191.4 NP_009122.2 2 - 3 664 66
GenBank mRNA AF122922.1 - 2 - 3 401 66
AK301818.1 - 2 - 3 591 66
AL711431.1 - 2 - 3 438 66
AY358344.1 - 2 - 3 434 66
BC018037.1 - 2 - 3 407 66
BX647427.1 - 2 - 3 333 66

View Full Product Details