The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243998.1 | NP_001230927.1 | 1 - 2 | 52 | 67 |
NM_001243999.1 | NP_001230928.1 | 1 - 2 | 52 | 67 | |
NM_001244000.1 | NP_001230929.1 | 1 - 2 | 52 | 67 | |
NM_003121.4 | NP_003112.2 | 1 - 2 | 52 | 67 | |
GenBank mRNA | AK225225.1 | - | 1 - 2 | 50 | 67 |
AK225720.1 | - | 1 - 2 | 52 | 67 | |
AK292644.1 | - | 1 - 2 | 54 | 67 | |
AK300639.1 | - | 1 - 2 | 50 | 67 | |
DA957263.1 | - | 1 - 2 | 49 | 67 |