The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077491.1 | NP_001070959.1 | 5 - 6 | 815 | 93 |
NM_001077492.1 | NP_001070960.1 | 4 - 5 | 747 | 93 | |
NM_012427.4 | NP_036559.1 | 4 - 5 | 950 | 93 | |
XM_011526702.1 | XP_011525004.1 | 4 - 5 | 689 | 93 | |
XM_011526703.2 | XP_011525005.1 | 5 - 6 | 769 | 93 | |
GenBank mRNA | AF168768.2 | - | 4 - 5 | 766 | 93 |
AF435980.1 | - | 4 - 5 | 921 | 93 | |
AF435981.1 | - | 4 - 5 | 718 | 93 | |
AK223421.1 | - | 4 - 5 | 898 | 93 | |
AY279380.1 | - | 4 - 5 | 724 | 93 | |
AY279381.1 | - | 5 - 6 | 792 | 93 | |
AY359010.1 | - | 4 - 5 | 950 | 93 | |
BC008036.2 | - | 4 - 5 | 881 | 93 | |
BT006867.1 | - | 3 - 4 | 597 | 93 |