Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSBG2
Entrez Gene ID
81616
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
BGR, BRGL, PRTD-NY3, PRTDNY3
Chromosome Location
Chr.19: 6135633 - 6193103 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289177.1 NP_001276106.1 6 - 7 910 104
NM_001289178.1 NP_001276107.1 6 - 7 856 104
NM_001289179.1 NP_001276108.1 6 - 7 901 104
NM_001289180.1 NP_001276109.1 6 - 7 620 104
NM_001321384.1 NP_001308313.1 6 - 7 891 104
NM_030924.4 NP_112186.3 6 - 7 901 104
XM_005259653.3 XP_005259710.1 6 - 7 883 104
XM_011528329.1 XP_011526631.1 2 - 3 188 104
XM_017027332.1 XP_016882821.1 6 - 7 728 104
XM_017027333.1 XP_016882822.1 6 - 7 883 104
XM_017027334.1 XP_016882823.1 6 - 7 883 104
GenBank mRNA AJ577571.1 - 6 - 7 804 104
AK093433.1 - 6 - 7 869 104
AK225862.1 - 5 - 6 609 104
AK302604.1 - 6 - 7 858 104
AL136854.1 - 6 - 7 642 104
AY009107.1 - 6 - 7 613 104
AY358766.1 - 6 - 7 901 104
BC022027.1 - 6 - 7 832 104
BC090046.1 - 6 - 7 817 104

View Full Product Details