Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location
Chr.19: 5205503 - 5340803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002850.3 NP_002841.3 11 - 12 1805 68
NM_130853.2 NP_570923.2 8 - 9 1766 68
NM_130854.2 NP_570924.2 8 - 9 1766 68
NM_130855.2 NP_570925.2 9 - 10 1778 68
XM_005259600.2 XP_005259657.1 8 - 9 1734 68
XM_005259606.2 XP_005259663.1 8 - 9 1734 68
XM_005259607.2 XP_005259664.1 8 - 9 1734 68
XM_005259609.1 XP_005259666.1 8 - 9 1733 68
XM_005259610.1 XP_005259667.1 8 - 9 1733 68
XM_011528157.2 XP_011526459.1 7 - 8 1604 68
XM_017027065.1 XP_016882554.1 9 - 10 1746 68
XM_017027066.1 XP_016882555.1 9 - 10 1764 68
XM_017027067.1 XP_016882556.1 9 - 10 1662 68
XM_017027068.1 XP_016882557.1 9 - 10 1746 68
XM_017027069.1 XP_016882558.1 9 - 10 1746 68
XM_017027070.1 XP_016882559.1 9 - 10 1746 68
XM_017027071.1 XP_016882560.1 9 - 10 1746 68
XM_017027072.1 XP_016882561.1 9 - 10 1746 68
XM_017027073.1 XP_016882562.1 3 - 4 949 68
XM_017027074.1 XP_016882563.1 9 - 10 1745 68
XM_017027075.1 XP_016882564.1 9 - 10 1745 68
XM_017027076.1 XP_016882565.1 9 - 10 1745 68
GenBank mRNA AB209333.1 - 8 - 9 1766 68
BC104812.1 - 7 - 8 1606 68
BC143286.1 - 7 - 8 1606 68
BC143287.1 - 7 - 8 1606 68
U35234.1 - 10 - 11 1679 68
U40317.1 - 7 - 8 1557 68
U41725.1 - 7 - 8 1557 68

View Full Product Details