Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLIP3
Entrez Gene ID
25999
Gene Name
CAP-Gly domain containing linker protein 3
Gene Aliases
CLIPR-59, CLIPR59, RSNL1
Chromosome Location
Chr.19: 36014660 - 36032895 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199570.1 NP_001186499.1 9 - 10 1641 87
NM_015526.2 NP_056341.1 10 - 11 1557 87
GenBank mRNA AJ427922.1 - 10 - 11 1535 87
AK056445.1 - 8 - 9 1040 87
AK090732.1 - 10 - 11 1560 87
AK092354.1 - 6 - 7 804 87
AK094738.1 - 10 - 11 1555 87
AK294748.1 - 10 - 11 1428 87
AL117468.1 - 2 - 3 169 87
BC013116.1 - 10 - 11 1469 87
BC014486.1 - 7 - 8 1000 87
BC128090.1 - 6 - 7 809 87

View Full Product Details