The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080722.4 | NP_542453.2 | 12 - 13 | 2222 | 56 |
NM_139155.3 | NP_631894.2 | 12 - 13 | 2231 | 56 | |
XM_011539300.2 | XP_011537602.1 | 12 - 13 | 2215 | 56 | |
XM_011539301.2 | XP_011537603.1 | 9 - 10 | 1228 | 56 | |
XM_011539302.2 | XP_011537604.1 | 9 - 10 | 1105 | 56 | |
XM_011539303.2 | XP_011537605.1 | 12 - 13 | 2219 | 56 | |
XM_011539305.2 | XP_011537607.1 | 12 - 13 | 2219 | 56 | |
XM_011539306.2 | XP_011537608.1 | 12 - 13 | 2219 | 56 | |
XM_011539307.2 | XP_011537609.1 | 12 - 13 | 2219 | 56 | |
XM_011539308.2 | XP_011537610.1 | 12 - 13 | 2219 | 56 | |
XM_011539309.1 | XP_011537611.1 | 5 - 6 | 815 | 56 | |
GenBank mRNA | AF358666.1 | - | 12 - 13 | 1925 | 56 |
AF366351.1 | - | 11 - 12 | 1852 | 56 | |
AJ345098.1 | - | 12 - 13 | 1925 | 56 | |
AK128469.1 | - | 5 - 6 | 815 | 56 |