The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278547.1 | NP_001265476.1 | 4 - 5 | 536 | 138 |
NM_001278548.1 | NP_001265477.1 | 3 - 4 | 353 | 138 | |
NM_001323302.1 | NP_001310231.1 | 6 - 7 | 799 | 138 | |
NM_001323320.1 | NP_001310249.1 | 4 - 5 | 536 | 138 | |
NM_001323321.1 | NP_001310250.1 | 4 - 5 | 536 | 138 | |
NM_001323322.1 | NP_001310251.1 | 6 - 7 | 799 | 138 | |
NM_001323323.1 | NP_001310252.1 | 5 - 6 | 605 | 138 | |
NM_001323324.1 | NP_001310253.1 | 4 - 5 | 536 | 138 | |
NM_001323325.1 | NP_001310254.1 | 5 - 6 | 730 | 138 | |
NM_001323326.1 | NP_001310255.1 | 5 - 6 | 605 | 138 | |
NM_001323327.1 | NP_001310256.1 | 5 - 6 | 730 | 138 | |
NM_001323328.1 | NP_001310257.1 | 5 - 6 | 605 | 138 | |
NM_001323329.1 | NP_001310258.1 | 4 - 5 | 536 | 138 | |
NM_001323330.1 | NP_001310259.1 | 5 - 6 | 730 | 138 | |
NM_001323331.1 | NP_001310260.1 | 5 - 6 | 730 | 138 | |
NM_139046.3 | NP_620634.1 | 6 - 7 | 799 | 138 | |
NM_139049.3 | NP_620637.1 | 6 - 7 | 799 | 138 | |
NR_136583.1 | 4 - 5 | 536 | 138 | ||
NR_136584.1 | 4 - 5 | 536 | 138 | ||
NR_136585.1 | 4 - 5 | 536 | 138 | ||
GenBank mRNA | AB451231.1 | - | 3 - 4 | 304 | 138 |
AB451271.1 | - | 3 - 4 | 304 | 138 | |
AB451354.1 | - | 3 - 4 | 304 | 138 | |
AB451398.1 | - | 3 - 4 | 304 | 138 | |
AK292523.1 | - | 4 - 5 | 432 | 138 | |
BC130570.1 | - | 3 - 4 | 366 | 138 | |
BC130572.1 | - | 3 - 4 | 366 | 138 | |
BC144063.1 | - | 3 - 4 | 366 | 138 | |
DA159335.1 | - | 4 - 5 | 536 | 138 | |
DQ234352.1 | - | 3 - 4 | 138 | ||
L26318.1 | - | 3 - 4 | 322 | 138 | |
U34822.1 | - | 3 - 4 | 316 | 138 | |
U35004.1 | - | 3 - 4 | 322 | 138 | |
U35005.1 | - | 3 - 4 | 317 | 138 |