The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190456.1 | NP_001177385.1 | 10 - 11 | 1233 | 120 |
NM_001190457.1 | NP_001177386.1 | 10 - 11 | 1512 | 120 | |
NM_001324014.1 | NP_001310943.1 | 11 - 12 | 1444 | 120 | |
NM_001324015.1 | NP_001310944.1 | 10 - 11 | 1245 | 120 | |
NM_006091.4 | NP_006082.3 | 10 - 11 | 1467 | 120 | |
GenBank mRNA | AB010098.1 | - | 10 - 11 | 1200 | 120 |
AB023142.1 | - | 9 - 10 | 1171 | 120 | |
AK124711.1 | - | 5 - 6 | 1687 | 120 | |
AK289678.1 | - | 10 - 11 | 1512 | 120 | |
BC026335.1 | - | 10 - 11 | 1183 | 120 |