Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAAA
Entrez Gene ID
27163
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
ASAHL, PLT
Chromosome Location
Chr.4: 75910655 - 75941013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014435.3 NP_055250.2 10 - 11 1173 135

View Full Product Details