Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
Chromosome Location
Chr.19: 8580240 - 8610735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030957.3 NP_112219.3 6 - 7 1119 73
XM_017027338.1 XP_016882827.1 5 - 6 975 73
GenBank mRNA AF163762.1 - 3 - 4 735 73

View Full Product Details