The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004260.3 | NP_004251.3 | 15 - 16 | 2800 | 60 |
XM_011517384.2 | XP_011515686.1 | 15 - 16 | 2602 | 60 | |
XM_017013992.1 | XP_016869481.1 | 14 - 15 | 2894 | 60 | |
XM_017013995.1 | XP_016869484.1 | 15 - 16 | 2802 | 60 | |
XM_017013998.1 | XP_016869487.1 | 14 - 15 | 2892 | 60 | |
GenBank mRNA | AB006532.1 | - | 15 - 16 | 2758 | 60 |
BC011602.2 | - | 8 - 9 | 1359 | 60 | |
BC013277.2 | - | 6 - 7 | 1097 | 60 | |
KJ147518.1 | - | 11 - 12 | 2107 | 60 |