The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248006.1 | NP_001234935.1 | 7 - 8 | 1922 | 61 |
NM_001248007.1 | NP_001234936.1 | 6 - 7 | 1585 | 61 | |
NM_006458.3 | NP_006449.2 | 8 - 9 | 1923 | 61 | |
NM_033278.3 | NP_150594.2 | 7 - 8 | 1758 | 61 | |
XM_011519844.1 | XP_011518146.1 | 8 - 9 | 2022 | 61 | |
XM_011519845.2 | XP_011518147.1 | 7 - 8 | 1703 | 61 | |
XM_017017098.1 | XP_016872587.1 | 7 - 8 | 1734 | 61 | |
XM_017017099.1 | XP_016872588.1 | 7 - 8 | 1626 | 61 | |
GenBank mRNA | AF045239.1 | - | 6 - 7 | 1527 | 61 |
AF220020.1 | - | 6 - 7 | 1527 | 61 | |
AF220021.1 | - | 7 - 8 | 1494 | 61 | |
AF220022.1 | - | 6 - 7 | 1290 | 61 | |
AK126685.1 | - | 5 - 6 | 1151 | 61 | |
AK291396.1 | - | 7 - 8 | 1922 | 61 | |
AK298843.1 | - | 6 - 7 | 1461 | 61 | |
AK299586.1 | - | 7 - 8 | 1715 | 61 | |
BC096827.1 | - | 7 - 8 | 1758 | 61 |