The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127892.1 | NP_001121364.1 | 2 - 3 | 3672 | 78 |
NM_002968.2 | NP_002959.2 | 2 - 3 | 3562 | 78 | |
XM_006721241.3 | XP_006721304.1 | 3 - 4 | 3637 | 78 | |
XM_011523254.2 | XP_011521556.1 | 3 - 4 | 3663 | 78 | |
GenBank mRNA | AK295103.1 | - | 3 - 4 | 3452 | 78 |
EU668343.1 | - | 2 - 3 | 3531 | 78 | |
Y18265.1 | - | 2 - 3 | 3531 | 78 |