Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANK1
Entrez Gene ID
53354
Gene Name
pantothenate kinase 1
Gene Aliases
PANK
Chromosome Location
Chr.10: 89579497 - 89645572 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138316.3 NP_612189.2 5 - 6 1051 113
NM_148977.2 NP_683878.1 6 - 7 2006 113
NM_148978.2 NP_683879.1 6 - 7 1228 113
XM_017016333.1 XP_016871822.1 7 - 8 1974 113
XM_017016334.1 XP_016871823.1 7 - 8 1936 113
XM_017016335.1 XP_016871824.1 7 - 8 2764 113
XM_017016336.1 XP_016871825.1 6 - 7 1153 113
XM_017016337.1 XP_016871826.1 7 - 8 2670 113
XM_017016338.1 XP_016871827.1 4 - 5 1370 113
GenBank mRNA AF355198.1 - 5 - 6 1035 113
AK125428.1 - 7 - 8 1585 113
AK127777.1 - 5 - 6 927 113
AY027661.1 - 6 - 7 2006 113
AY027662.1 - 6 - 7 1228 113
AY027663.1 - 7 - 8 1322 113
BC026296.1 - 3 - 4 361 113

View Full Product Details