Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPFIA2
- Entrez Gene ID
-
8499
- Gene Name
- PTPRF interacting protein alpha 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 81257975 - 81759748 on Build GRCh38
View Full Product Details