The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304430.1 | NP_001291359.1 | 5 - 6 | 669 | 107 |
NM_014305.3 | NP_055120.1 | 5 - 6 | 593 | 107 | |
NR_130731.1 | 5 - 6 | 589 | 107 | ||
NR_130732.1 | 4 - 5 | 502 | 107 | ||
XM_011521065.2 | XP_011519367.1 | 5 - 6 | 596 | 107 | |
XM_011521066.2 | XP_011519368.1 | 5 - 6 | 477 | 107 | |
GenBank mRNA | AF048686.1 | - | 5 - 6 | 495 | 107 |
AJ006068.1 | - | 5 - 6 | 547 | 107 | |
AK303439.1 | - | 4 - 5 | 474 | 107 | |
AK313251.1 | - | 5 - 6 | 503 | 107 | |
AL527626.3 | - | 5 - 6 | 592 | 107 | |
AY544125.1 | - | 5 - 6 | 470 | 107 | |
BC005284.1 | - | 5 - 6 | 575 | 107 | |
BC033675.1 | - | 5 - 6 | 522 | 107 | |
BG546428.1 | - | 4 - 5 | 380 | 107 | |
BG702561.1 | - | 5 - 6 | 479 | 108 | |
BI914642.1 | - | 5 - 6 | 636 | 107 |