Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA5A
Entrez Gene ID
9037
Gene Name
semaphorin 5A
Gene Aliases
SEMAF, semF
Chromosome Location
Chr.5: 9035026 - 9546121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003966.2 NP_003957.2 12 - 13 2195 56
XM_006714506.2 XP_006714569.1 13 - 14 2294 56
XM_006714507.3 XP_006714570.1 11 - 12 1857 56
XM_011514155.2 XP_011512457.1 12 - 13 1818 56
XM_011514156.1 XP_011512458.1 14 - 15 2367 56
XM_011514157.1 XP_011512459.1 13 - 14 2222 56
XM_011514158.1 XP_011512460.1 11 - 12 2052 56
XM_011514159.2 XP_011512461.1 8 - 9 4814 56
XM_017010016.1 XP_016865505.1 11 - 12 2049 56
GenBank mRNA AK293441.1 - 2 - 3 382 56
AK307587.1 - 12 - 13 2149 56
BC115696.1 - 12 - 13 1927 56
KJ534937.1 - 10 - 11 1483 56
U52840.1 - 12 - 13 2120 56

View Full Product Details