Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA5A
Entrez Gene ID
9037
Gene Name
semaphorin 5A
Gene Aliases
SEMAF, semF
Chromosome Location
Chr.5: 9035026 - 9546121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003966.2 NP_003957.2 21 - 22 3602 148
XM_006714506.2 XP_006714569.1 22 - 23 3701 148
XM_006714507.3 XP_006714570.1 20 - 21 3264 148
XM_011514155.2 XP_011512457.1 21 - 22 3225 148
XM_011514156.1 XP_011512458.1 23 - 24 3774 148
XM_011514157.1 XP_011512459.1 22 - 23 3629 148
XM_011514158.1 XP_011512460.1 20 - 21 3459 148
XM_011514159.2 XP_011512461.1 17 - 18 6221 148
XM_017010016.1 XP_016865505.1 20 - 21 3456 148
GenBank mRNA AK293441.1 - 11 - 12 1789 148
AK307587.1 - 21 - 22 3556 148
BC115696.1 - 21 - 22 3334 148
KJ534937.1 - 19 - 20 2890 148
KJ535077.1 - 18 - 19 2682 148
U52840.1 - 21 - 22 3527 148

View Full Product Details