Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA5A
Entrez Gene ID
9037
Gene Name
semaphorin 5A
Gene Aliases
SEMAF, semF
Chromosome Location
Chr.5: 9035026 - 9546121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003966.2 NP_003957.2 8 - 9 1358 116
XM_006714506.2 XP_006714569.1 9 - 10 1457 116
XM_006714507.3 XP_006714570.1 7 - 8 1020 116
XM_011514155.2 XP_011512457.1 8 - 9 981 116
XM_011514156.1 XP_011512458.1 10 - 11 1530 116
XM_011514157.1 XP_011512459.1 9 - 10 1385 116
XM_011514158.1 XP_011512460.1 7 - 8 1215 116
XM_011514159.2 XP_011512461.1 4 - 5 3977 116
XM_017010016.1 XP_016865505.1 7 - 8 1212 116
GenBank mRNA AK307587.1 - 8 - 9 1312 116
BC115696.1 - 8 - 9 1090 116
KJ534937.1 - 6 - 7 646 116
KJ535077.1 - 6 - 7 646 116
U52840.1 - 8 - 9 1283 116

View Full Product Details