The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077261.3 | NP_001070729.2 | 35 - 36 | 5480 | 70 |
NM_001206654.1 | NP_001193583.1 | 35 - 36 | 5480 | 70 | |
NM_006312.5 | NP_006303.4 | 36 - 37 | 5510 | 70 | |
GenBank mRNA | AF113003.1 | - | 35 - 36 | 5190 | 70 |
AF125672.1 | - | 34 - 35 | 5315 | 70 | |
AY965853.1 | - | 34 - 35 | 5315 | 70 | |
BC020427.1 | - | 1 - 2 | 226 | 70 | |
S83390.1 | - | 2 - 3 | 538 | 70 |