The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101339.2 | NP_001094809.2 | 3 - 4 | 616 | 94 |
XM_017018966.1 | XP_016874455.1 | 4 - 5 | 627 | 94 | |
GenBank mRNA | BC035880.1 | - | 3 - 4 | 627 | 94 |