The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198173.2 | NP_937816.1 | 1 - 2 | 248 | 68 |
NM_198174.2 | NP_937817.3 | 1 - 2 | 248 | 68 | |
GenBank mRNA | AK074386.1 | - | 1 - 2 | 145 | 68 |
AK315164.1 | - | 1 - 2 | 50 | 68 | |
BU848928.1 | - | 1 - 2 | 179 | 68 | |
DA714132.1 | - | 1 - 2 | 248 | 68 |